VWR®, Flasks, Round Bottom, Narrow or Wide Neck
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
214-1182EA 8.3 GBP
214-1182 214-1177 214-1178 214-1179 214-1180 214-1181 214-1183 214-1184 214-1185 214-1186
VWR®, Flasks, Round Bottom, Narrow or Wide Neck
Flasks Round Bottom Flasks
Borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The economical alternative for a wide range of applications. Marked with capacity and a white labelling area.
ISO 1773 (narrow mouth)
DIN 12347 (wide mouth)
Certifications: ISO 1773 (narrow neck)
DIN 12347 (wide neck)