Distillation flasks, engler type, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
216532807 216532901
752-0500EA 44.8 GBP
752-0500 214-0435
Distillation flasks, engler type, DURAN®
Flasks Distillation Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Uniform wall thickness distribution makes these flasks ideal for heating applications and distillations.