Erlenmeyer flasks, narrow neck with strengthened rim
Supplier: DIXON SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
236006503 236006504 236006505 236006507 236006508 236006510
214-0160EA 66.3 GBP
214-0160 214-0161 214-0162 214-0163 214-0164 214-0165
Erlenmeyer flasks, narrow neck with strengthened rim
Flasks Erlenmeyer Flasks
Borosilicate glass, grade 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Reduced breakage
For a wide range of laboratory applications. Volume graduations and labelling area.
Order Now
SPECIFICATIONS
Environmentally Preferable