Beakers, low form
Supplier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
110205 110305 110405 110605 110905 111005
213-1602EA 44.9 GBP
213-1602 213-1603 213-1604 213-1605 213-1606 213-1609
Beakers, low form
Beakers
PFA, translucent, with spout.
  • Excellent chemical resistance
  • High thermal resistance from –270 to +250 °C
  • Autoclavable
Ideal for demanding applications. Not for use on hotplates. With engraved graduation.
ISO 7056
Certifications: ISO 7056