Ammonium heptamolybdate tetrahydrate ≥98.5%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Hexaammonium heptamolybdate tetrahydrate, Ammonium molybdate(VI) tetrahydrate, Ammonium molybdate tetrahydrate, Hexammonium heptamolybdate 4-hydrate, Molybdic acid ammonium salt tetrahydrate

21275.262EA 341 GBP
21275.262
Ammonium heptamolybdate tetrahydrate ≥98.5%, GPR RECTAPUR®
Ammonium heptamolybdate tetrahydrate
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now

Specification Test Results

Assay Min. 98.5 %
Heavy metals (as Pb) Max. 50 ppm
PO4 + AsO4 + SiO4 (as PO4) Max. 0.02 %
Fe (Iron) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR