Ammonium chloride ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Ammonium muriate, Salmiac

21235.297EA 70.5 GBP
21235.297 21235.366 21235.468
Ammonium chloride ≥99%, GPR RECTAPUR®
Ammonium chloride
Formula: NH₄Cl
Melting Pt: 335 °C
Density: 1.5256 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
REACH: 01-2119487950-27
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.9 %
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR