Beakers, triangular, Tricorn™
Supplier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B700-100 B700-1L B700-250 B700-400 B700-800 B700-50
212-9302EA 78.5 GBP
212-9302 212-9306 212-9303 212-9304 212-9305 212-9301
Beakers, triangular, Tricorn™
Beakers
These Tricorn™ beakers are made of polypropylene, provide three dripless pouring spouts that offers more security.
  • Autoclavable
  • Tough and unbreakable
Suitable for use with commonly used acids, alkalis and solvents.
Packaging: All sizes are available 100 per case.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904