Test tubes with rim, light wall
Supplier: MURRAY S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
G001/82 G001/71 G001/72 G001/73 G001/75 G001/76 G001/80 G001/81 G001/83 G001/84 G001/79
212-0556EA 120 GBP
212-0556 212-7076 212-7077 212-7078 212-7079 212-7080 212-7081 212-7082 212-7083 212-7084 212-0555
Test tubes with rim, light wall
Tubes Test Tubes
Soda-lime glass test tubes are suitable for general mixing and simple laboratory work, and will withstand moderate warming (water bath).
Caution: They should not be placed directly into naked flames.
Order Now
SPECIFICATIONS
Environmentally Preferable