Sample vials, trident vials, squat
Supplier: MURRAY S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
T103/V1T003/C T103/V2T003/C T103/V3T003/C T103/V5T003/C
212-7063EA 185 GBP
212-7063 212-7065 212-7067 212-7070
Sample vials, trident vials, squat
Vials
Vials made from neutral glass with snap-on lid. Designed for a wide variety of uses in the laboratory, production, more.
  • Wide opening for easy filling and emptying
  • With beaded rim and sealing snap-on lid
  • Robust and economical, will withstand moderate temperature change
Packaging: For production related reasons, the number of vials per box may vary.