VWR®, Test Tubes with Rim
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
212-0753EA 26.7 GBP
212-0753 212-0754 212-0755 212-0756 212-0757 212-0758 212-0759 212-0760 212-0761 212-0762 212-0763 212-0764 212-0765
VWR®, Test Tubes with Rim
Tubes Test Tubes
Test tubes made from borosilicate 5.1 glass, neutral glass.
  • Thin walled
  • Excellent chemical resistance
  • Relatively resistant to rapid temperature changes and local heating
The economical alternative for a wide range of uses.
Certifications: DIN 12395.
Order Now
SPECIFICATIONS
Environmentally Preferable