Gas distribution tubes, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
258561007 258562106 258561101 258560208 258561204 258560002 258560105
212-0675EA 274 GBP
212-0675 212-0678 212-0676 212-0674 212-0677 212-0672 212-0673
Gas distribution tubes, DURAN®
Gas Tubes
These gas distribution tubes are designed for reactions between gases and liquids. They are made of clear DURAN® borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Order Now
SPECIFICATIONS
Environmentally Preferable