Test tube racks, Azlon®
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
RPW013P RPW018P RPW021P RPW026P RPW031P RPB018P RPB026P RPB031P RPB021P
212-0081EA 0 GBP
212-0081 212-0082 212-0083 212-0084 212-0085 AZLORPB018P AZLORPB026P AZLORPB031P AZLORPB021P
Test tube racks, Azlon®
Racks Tube Racks
PP
  • Wide range of sizes to fit tubes from 13 to 31 mm diameter
  • Autoclavable and the racks sink in water baths
  • Alpha-numeric place coding
Sturdy test tube racks supplied in self-assembly flat packs.