Draining racks, wall mounted
Supplier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1557 2110
211-2084EA 446 GBP
211-2084 SEMA2110
Draining racks, wall mounted
Racks Draining Racks
White, PVC drain channel, polyethylene coated metal rack.
  • Drain channels inclined toward the centre and provided with a drain point
  • Metal frame and drip plasticised PE rods
  • The rack is hung on the wall plate and can be easily removed for cleaning