VWR®, Racks for Centrifuge Tubes and Bottles
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
525-0741EA 38.6 GBP
525-0741 211-0197 525-0742
VWR®, Racks for Centrifuge Tubes and Bottles
Racks Tube Racks
Steel wire with an epoxy coating.
  • Ribbed base for better tube stability
  • Very good chemical resistance
Racks for 15 and 50 ml centrifuge tubes and 250 ml centrifuge bottles. Can also be used for all other types of tubes.
Order Now
SPECIFICATIONS
Environmentally Preferable