Dulbecco's phosphate-buffered saline (DPBS), Corning®

Supplier: Corning
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

21-031-CM 55-031-PB 55-031-PC 20-030-CV 20-031-CV 21-030-CV 21-031-LX 21-031-LB 21-031-CV 21-031-CM 21-030-CM
MDTH21-031-CMEA 107 GBP
MDTH21-031-CM 55-031-PB 55-031-PC 20-030-CV 20-031-CV 21-030-CV 21-031-LX 21-031-LB 21-031-CV 21-031-CM 21-030-CM
Dulbecco's phosphate-buffered saline (DPBS), Corning®
Buffers Cell Culture Buffers

Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7,0 to 7,6


  • Low endotoxin - < 0,25 EU/ml
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment


It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.


Certifications: Complete Certificate of Analysis available for each production lot, along with full formulation information.


Caution: Storage temperature: Solutions 2 - 30 °C; Powder formats 2 - 8 °C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR