Hank's balanced salt solutions (HBSS), Corning®
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20-023-CV 21-020-CM 21-020-CV 21-021-CM 20-021-CVR 21-021-CV 21-023-CM 21-022-CV 21-023-CV 21-022-CM
20-023-CVEA 290 GBP
20-023-CV 21-020-CM 21-020-CV 21-021-CM MDTC20-021-CVR 21-021-CV 21-023-CM 21-022-CV 21-023-CV 21-022-CM
Hank's balanced salt solutions (HBSS), Corning®
Buffers Cell Culture Buffers
Hanks' Balanced Salt Solution (HBSS) is a buffer used to maintain a physiological pH for cells maintained in non-CO₂ atmospheric conditions.
  • Low endotoxin - < 0.25 EU/ml
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment