Formaldehyde 36% aqueous solution Stabilised, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilisation: 10 % MeOH

20910.294EA 44.6 GBP
20910.294 20910.328 20910.363 20910.443 20910.330
Formaldehyde 36% aqueous solution Stabilised, GPR RECTAPUR®
Formaldehyde
Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
Density: 1.088 g/cm³ (20 °C)
Flash Pt: 66 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 2209
ADR: 8 III
Merck Index: 13,04259

Order Now

Specification Test Results

Assay 35 - 38 %
Density (20/4) 1.083 - 1.093
Heavy metals (as Pb) Max. 20 ppm
Ignition residue (SO4) Max. 0.02 %
Methanol 8 - 12 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR