Formaldehyde 36% in aqueous solution stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilisation: Stabilised with methanol ∼10 %

20909.290EA 61.1 GBP
20909.290 20909.368 20909.448 20909.330 20909.550 20909.325
Formaldehyde 36% in aqueous solution stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Formaldehyde
Formula: CH₂O
Boiling Pt: 100 °C
Density: 1.088 g/cm³ (20 °C)
Flash Pt: 66 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 2209
ADR: 8,III

Order Now

Specification Test Results

Assay 36.5 - 38.0 %
Acidity Max. 0.006 meq/g
Appearance of solution Passes test Ph.Eur.
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
Identification C Passes test Ph.Eur.
IR Spectrum Passes test
Colouration Max. 10 APHA
Density (20/4) 1.083 - 1.093
Heavy metals (as Pb) Max. 5 ppm
Ignition residue (SO4) Max. 50 ppm
Methanol 8 - 10 %
Methanol (V/V) 9.0 - 15.0 %
Cl (Chloride) Max. 5 ppm
SO4 (Sulphate) Max. 20 ppm
Fe (Iron) Max. 2 ppm
Pb (Lead) (ISO) Max. 2 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR