Vydac 208TP C8, HPLC columns
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
554-7581EA 1030 GBP
554-7581 208TP510 208TP3405 208TP3310 208TP3410 208GCC510 208FSK510 208TP5105 208TP5205 208TP5210 208TP5115 208TP5215 208TP5415 208TP52 208TP53 208TP104 208TP1010 208TP1022 208GCC1210 208FSK1210 208TP101510 208TP101522 208TP152022 208GD104/N 208GD34/N 208GD52/N 208GD54/N 208GK104/N 208GK34/N 208GK52/N 208GK54/N 208GCC1010 208TP3415
Vydac 208TP C8, HPLC columns
Chromatography Columns
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
208TP C8 is less hydrophobic than C18TP and is recommended for the analysis of peptides, 10 – 20KMore Product Information