Amidosulphonic acid ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

20675.290EA 48 GBP
20675.290 20675.320 20675.460
Amidosulphonic acid ≥99%, GPR RECTAPUR®
Amidosulphonic acid
Formula: H₃NO₃S
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 205 °C
Density: 2.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: 8,III
Merck Index: 13,09006
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue Max. 0.05 %
Water Max. 0.1 %
SO4 (Sulphate) Max. 0.1 %
Fe (Iron) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR