Anti-IgY Goat Polyclonal Antibody (CF™ 660C)

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

20371-50UL
20371-50ULEA 144 GBP
20371-50UL
Anti-IgY Goat Polyclonal Antibody (CF™ 660C)
Antibodies
CF™660C and CF™660R antibodies are affinity-purified antibodies labeled with far-red fluorescent CF™660C or CF™660R dyes, members of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.

CF™660C is a far-red fluorescent dye with excitation/emission maxima at 667/685nm. CF™660C is several fold brighter and more photostable than the spectrallyMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol IgY
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation CF® 660C
Reactivity Chicken
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Flow cytometry Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR