Anti-IgG Chicken Polyclonal Antibody (CF™ 568)

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

20338-50UL
20338-50ULEA 115 GBP
20338-50UL
Anti-IgG Chicken Polyclonal Antibody (CF™ 568)
Antibodies
CF™568 antibodies are affinity-purified antibodies labeled with red fluorescent CF™568, a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.

CF™568 is a red fluorescent dye optimized for the 568nm laser line. CF™568 is spectrally similar to AlexaFluor®568, ATTO 565, and Rhodamine Red. Antibody conjugates of CF™568 areMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation CF® 568
Reactivity Mouse
Host Chicken
Isotype IgY
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Flow cytometry Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR