Hydrochloric acid 25%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

20257.296EA 38.4 GBP
20257.296 20257.321 20257.310 20257.401 20257.460
Hydrochloric acid 25%, AnalaR NORMAPUR® analytical reagent
Hydrochloric acid
Formula: HCl
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
REACH: 01-2119484862-27

Order Now

Specification Test Results

Assay Min. 25.0 %
Colouration Max. 10 APHA
Free chlorine Max. 1 ppm
Ignition residue (SO4) Max. 5 ppm
Br (Bromide) Max. 50 ppm
NH4 (Ammonium) Max. 1 ppm
SO3 (Sulphite) Max. 1 ppm
SO4 (Sulphate) Max. 2 ppm
Al (Aluminium) Max. 0.2 ppm
As (Arsenic) Max. 0.01 ppm
Ba (Barium) Max. 0.05 ppm
Be (Beryllium) Max. 0.02 ppm
Bi (Bismuth) Max. 0.1 ppm
Ca (Calcium) Max. 1 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.2 ppm
Ge (Germanium) Max. 0.05 ppm
K (Potassium) Max. 0.1 ppm
Li (Lithium) Max. 0.01 ppm
Mg (Magnesium) Max. 0.2 ppm
Mn (Manganese) Max. 0.01 ppm
Mo (Molybdenum) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.02 ppm
Sr (Strontium) Max. 0.01 ppm
Ti (Titanium) Max. 0.1 ppm
Tl (Thallium) Max. 0.05 ppm
V (Vanadium) Max. 0.01 ppm
Zn (Zinc) Max. 0.1 ppm
Zr (Zirconium) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR