Hydrochloric acid 30%, TECHNICAL

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

20251.365EA 76.3 GBP
20251.365 20251.445
Hydrochloric acid 30%, TECHNICAL
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
REACH: 01-2119484862-27

Order Now

Specification Test Results

Assay 29 - 31 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR