Distilling bridges, Quickfit®
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SH2/33
201-8692EA 46.3 GBP
201-8692
Distilling bridges, Quickfit®
Distillation Components Distillation Bridges
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
Used in destillation assemblies in which the condenser is assembled in the vertical mode.