Multiple adapters, Quickfit®
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MA1/2 MA1/3 MA1/11 MA1/33
201-8572EA 50.6 GBP
201-8572 201-8573 201-8571 201-8574
Multiple adapters, Quickfit®
Distillation Components Multiple Distillation Adapters
PYREX®, borosilicate glass, straight.
  • Both sockets are the same size
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters with two parallel necks/sockets and a cone.