Rotary evaporating flasks
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FD50/4RE FD250/4RE FD500/4RE FD1L/4RE FD2L/4RE FD250/3 FD100/3 FD100/4RE FD500/3 FD1L/3
201-8404EA 29.6 GBP
201-8404 201-8403 201-8406 201-8401 201-8009 201-8402 531-0741 201-2374 BIBBFD500/3 BIBBFD1L/3
Rotary evaporating flasks
Flasks Rotary Evaporator Flasks
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for use with rotary evaporators. Large surface area for rapid evaporation. Robust heavy wall construction for added strength. Florentine shape allows easy removal of solid residues.