Dropping funnels, cylindrical, with pressure balance
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
04372811 04373711 04374914 04375814 04377014 04372814 04373714 04372211
201-5172EA 155 GBP
201-5172 201-5173 201-5184 201-5185 201-5186 201-5182 201-5183 201-0897
Dropping funnels, cylindrical, with pressure balance
Funnels Dropping Funnels
DURAN®, borosilicate glass, clear, with PTFE key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint cone and socket, with divisions for approximate volume reading. Stopcock with thread guard.