Distilling bridges
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05053304 05053404 05053201 05053301 05053311 05053211 05053314 05053414
201-3131EA 254 GBP
201-3131 201-3132 201-0899 201-1067 LENZ05053311 LENZ05053211 LENZ05053314 LENZ05053414
Distilling bridges
Distillation Components Distillation Bridges
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Bridge includes fused Liebig condenser and curved vacuum adapter. NS 14/23 socket for distillation thermometer with 55 mm fitted length. Water and vacuum connection via hose connectors with 8 mm external diameter or water connection with thread GL 14 and vacuum receiver adapter with thread GL 18.