Gas washing bottles, with head, VitraPOR®
Supplier: ROBU GLASFILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
34251 34101 34252 34250 34100 34102
201-1460EA 258 GBP
201-1460 201-1461 ROBU34252 ROBU34250 ROBU34100 ROBU34102
Gas washing bottles, with head, VitraPOR®
Bottles Gas Washing Bottles
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Head with fritted disc and standard taper joint.