VWR® Reduction Adapters for Conical Ground Joints
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
212-0858EA 10.2 GBP
212-0858 212-0859 212-0860 212-0861 212-0862 212-0863 212-0864 201-1436 201-1437 201-1438
VWR® Reduction Adapters for Conical Ground Joints
Adapters Adapters, Conical Ground Joints
Adapters from large cone at the top to small socket at the bottom.
  • Made from clear borosilicate 3.3 glass
  • Excellent chemical resistance, high temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Outstanding leak tightness
Certifications: Produced according to DIN 12257.