VWR® Splash Heads, Stutzer, Straight
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
201-1398EA 44.8 GBP
201-1398 201-1399 201-1400 201-1403
VWR® Splash Heads, Stutzer, Straight
Distillation Components Distillation Heads
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • Straight, with standard ground joint and socket
These splash heads, according to Stutzer, can be placed directly onto the distillation flask to intercept droplets.