VWR® Erlenmeyer Flasks, with Standard Ground Joint
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
201-1385EA 20.3 GBP
201-1385 201-1386 201-1387 201-1388 201-1389 201-1390 201-1391 201-1392 201-1393 201-1394 201-1395 201-1372 201-1373 201-1362 201-1363 201-1369 201-1370
VWR® Erlenmeyer Flasks, with Standard Ground Joint
Flasks Erlenmeyer Flasks
Borosilicate 3.3 glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Good value alternative for a wide range of laboratory applications. Volume graduations and labelling area.
DIN 12387
Certifications: DIN 12387