Flasks, round bottom, with standard ground joint
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03001413 03001422 03001428 03001437 03001449 03001922 03001928 03001937 03001949 03001958 03002428 03002437 03002449 03002458 03002470 03002473 03002928 03002937 03002949 03002958 03002970 03002973 03002976 03002979 03002982 03002985 03002988 03003470 03004549 03004558 03004570 03004573 03004576 03004579 03004585 03004588 03004582 03001410 03002994 03004594
201-1301EA 14.3 GBP
201-1301 201-1302 201-1303 201-1304 201-1305 201-1310 201-0097 201-0098 201-0099 201-0100 271-1306 271-1307 271-1305 271-1308 271-1309 271-1310 201-1323 201-1324 201-1325 201-1329 201-1327 201-1328 201-1332 201-1331 201-1330 201-0101 201-0102 201-0103 201-1345 201-1346 201-1347 271-1348 LENZ03004576 201-1349 271-1351 271-1352 271-1350 201-1195 LENZ03002994 LENZ03004594
Flasks, round bottom, with standard ground joint
Flasks Round Bottom Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
DIN 12348
Certifications: DIN EN ISO 4797
Order Now
SPECIFICATIONS
Environmentally Preferable