VWR® Dropping funnels, cylindrical, without pressure balance
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
201-1238EA 58.2 GBP
201-1238 201-1239 201-1240 201-1241 201-1242 201-1243 201-1244
VWR® Dropping funnels, cylindrical, without pressure balance
Funnels Dropping Funnels
Borosilicate 3.3 glass.
  • Ground glass cone for flasks and assemblies
  • PTFE key stopcock and PP stopper
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
PTFE key stopcock for non stick, grease-free performance.
Certifications: ISO 4800
Order Now
SPECIFICATIONS
Environmentally Preferable