Joint clamps for conical ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
01103712 01103719 01103724 01103734 01103745 01103714 01103729
201-0765EA 82.5 GBP
201-0765 271-1121 271-1122 271-1124 201-0767 201-0900 201-0901
Joint clamps for conical ground joints
Clamps NS Joint Clamps
POM, coloured. For firm, securely fitting conical ground joints. These clamps are made of plastic to avoid scratching the glass, as happens with conventional metal joint clamps.
  • Very good chemical resistance
  • Heat resistant up to +150 °C
  • Sturdy, easy to install and remove