Powder flasks with conical ground joint
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03043070 03043073 03043037 03043049 03043058
201-0628EA 281 GBP
201-0628 LENZ03043073 LENZ03043037 201-0626 201-0627
Powder flasks with conical ground joint
Flasks Rotary Evaporator Flasks
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for rotary evaporators, for drying of powder-like substances. With center neck, conical joint and inscription label.