Flasks, round bottom with three necks and standard ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03311137 03313237 03313249 03313258 03313270 03313273 03314137 03314149 03314158 03314170 03314173 03314179 03314237 03314249 03314258 03314270 03314437 03314449 03314458 03314470 03314473 03314476 03314479 03314488 03313149 03311128 03315470 03315473 03313137 03315476 03315479 03315482 03315485 03315488 03315494 03314485 03313170 03313158 03314482 03311122 03314176 03314185 03314273
271-1514EA 94 GBP
271-1514 201-1178 201-1179 271-1512 201-1180 201-1181 201-1501 201-1502 201-1504 201-1506 271-1509 201-1051 201-1053 201-1054 201-1055 201-1056 201-0336 201-1503 201-1505 201-1507 201-1510 201-1511 201-1058 201-1182 271-1513 201-1176 271-1508 271-1511 201-1177 201-1061 201-1062 201-1063 201-1064 201-1065 201-1183 201-1060 201-1196 201-1049 201-1059 201-0613 201-1050 201-1052 201-1057
Flasks, round bottom with three necks and standard ground joints
Flasks Round Bottom Flasks
DURAN®, borosilicate glass, with angled side necks, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck and two side necks, both inclined at an angle of 20°. Matt white labelling area.