Splash heads, anti-climb, Quickfit®
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SH15/29 SH15/24 SH15/19
201-0573EA 111 GBP
201-0573 201-0578 BIBBSH15/19
Splash heads, anti-climb, Quickfit®
Distillation Components Distillation Heads
PYREX® borosilicate glass.
  • Protects against liquid ingress into the condenser
  • Useful for liquids that have tendency to foam
  • Splash heads feature a 29/32 socket
For use with rotary evaporators.