Fractionating columns, Vigreux
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.521.200 1.522.300 1.523.600 1.521.300 1.523.300 1.521.201 1.520.201 1.523.201
201-0175EA 124 GBP
201-0175 201-0177 201-0179 201-0176 201-0178 WITG1.521.201 WITG1.520.201 WITG1.523.201
Fractionating columns, Vigreux
Distillation Components Distillation Columns
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With or without glass jacket.