Gas washing bottles, PFA, PTFE frit and tube connectors, BOLA

Supplier: Bohlender
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A118-01 A118-02 A118-03
201-0015EA 605 GBP
201-0015 201-0016 201-0017
Gas washing bottles, PFA, PTFE frit and tube connectors, BOLA
Bottles Gas Washing Bottles
Transparent PFA bottle with white PTFE frit.

  • Pore-free, resistant to practically all chemicals
  • Fine-pore frit of 3 µm ensures optimal pearling of the gas to be washed
  • Temperature resistance from −200 to +250 °C

Stable, robust bottles.
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR