Aluminium oxide activated neutral Brockmann Grade I ~150 mesh, Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

199974-1KG 199974-5KG 199974-25KG
199974-1KGEA 121 GBP
199974-1KG 199974-5KG HONC199974-25KG
Aluminium oxide activated neutral Brockmann Grade I ~150 mesh, Fluka™
Aluminium oxide activated neutral
Formula: Al₂O₃
Boiling Pt: 2.980 °C (1.013 hPa)
Melting Pt: 2.072 °C
Density: 3.960 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Specification Test Results

Brockmann Number (Activity) I (val.ref) complying
pH (10 % susp., 20°C) 6.5 - 7.5
Spez. surface (BET) 135 - 165 m²/g
Fe (as Fe₂O₃) Max. 0.03 %
Na (as Na₂O) Max. 0.4 %
Silicon (as SiO₂) Max. 0.03 %
Chloride (Cl) Max. 0.2 %
Grain size 0.04-0.30 mm Min. 95 %
Pore Size 58 +� pore size


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR