Potassium carbonate, anhydrous 99+% for analysis

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Potash

198040010 198040050 198040250
198040010.EA 54 GBP
198040010. 198040050. 198040250.
Potassium carbonate, anhydrous 99+% for analysis
Potassium carbonate
Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891 °C
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder or granular powder
Loss on drying ≤1 % (300°C)
Titration with HClO4 ≥99.0 % (On dried substance)
Chloride (Cl) ≤30 ppm
Phosphate (PO4) ≤50 ppm
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤10 ppm
Sodium (Na) ≤1000 ppm
Heavy metals (as Pb) ≤5 ppm
Total sulfur (as SO4) ≤0.003 %
Total nitrogen (as N) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR