ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), Sigma-Aldrich®

Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

194430-1
194430-1EA 127 GBP
194430-1
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), Sigma-Aldrich®
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])

ABTS™ Chromophore, Diammonium Salt


Useful as a substrate for ELISA; acts as a hydrogen donor in peroxidase reactions. Radical cation has absorption maximum at 414 nm.

Formula: C₁₈H₂₄N₆O₆S₄
Storage Temperature: Refrigerator
MDL Number: MFCD00010404
CAS Number: 30931-67-0
EINECS: 250-396-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR