Toluene ≥99.7% for spectrophotometry

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

19376.K2 19376.M1
19376.K2EA 36.7 GBP
19376.K2 19376.M1
Toluene ≥99.7% for spectrophotometry
Toluene

Purity: 99.7+% CAS No.: 108-88-3 Molecular Formula: C6H5CH3 Formula Weight: 92.14 Flash Point: 4°(40°F) Physical Form: Liquid Density: 0.867 Refractive Index: 1.4967 Appearance: Colourless Odour: Aromatic Melting Point: -93° Boiling Point: 111° Beilstein Reference: 635760 Merck Reference: 14,9529 UN No.: 1294 MDL No.: MFCD00008512

Formula: C₆H₅CH₃
Boiling Pt: 111 °C
Melting Pt: –93 °C
Density: 0.867
Flash Pt: 4 °C (40 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 14,9529
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR