Dextrose, anhydrous 97,5-102,0 Anhydrous basis USP, multi-compendial, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

1920-05EA 272 GBP
1920-05 1920-07 1920-06 1920-09 1920-20 1920-37 1920-39 1920-01
Dextrose, anhydrous 97,5-102,0 Anhydrous basis USP, multi-compendial, J.T.Baker®
D(+)-Glucose

Dextrose multi-compendial.
Make upstream processing steps more reliable and productive with pharmaceutical sugars from Avantor™ Performance Materials. These products are a reliable source of carbon and energy for cell cultures, and used as excipients, effectively stabilize valuable and delicate lyophilized proteins for injectable biopharmaceutical drugs.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets J.P. Chemical Specifications
Meets U.S.P Requirements
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Preserve in well-closed containers.
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C: Water (H₂O) ≤1.0 %
USP - Assay (Anhydrous basis) 97.5 - 102.0 %
USP - Related substances: Maltose and isomaltose ≤0.4 %
USP - Related substances: Maltotriose ≤0.2 %
USP - Related substances: Fructose ≤0.15 %
USP - Related substances: Unspecified impurities, each ≤0.10 %
USP - Related substances: Total Impurities ≤0.5 %
USP - Color and Clarity of Solution Passes Test
USP - Dextrin Passes Test
USP - Soluble Starch, Sulfites ≤15 ppm
USP - Conductivity (uS cm-1) ≤20 µS/cm
USP - Endotoxin Concentration (EU/g) ≤2.5
EP/BP - Identification A (Specific Rotation (+)) 52.5 - 53.3 Degree
EP/BP - Identification B Passes Test
EP/BP - Identification E Passes Test
EP/BP - Assay (C₆H₁₂O₆) (anhydrous substance) 97.5 - 102.0 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Conductivity, uS cm-1 ≤20
EP/BP - Related Substances: sum of impurities A and B ≤0.4 %
EP/BP - Related Substances: impurity C ≤0.2 %
EP/BP - Related Substances: impurity D ≤0.15 %
EP/BP - Related Substances: unspecified impurities, each ≤0.10 %
EP/BP - Related Substances: total impurities ≤0.5 %
EP/BP - Dextrins Passes Test
EP/BP - Soluble starch, sulfite ≤15 ppm
EP/BP - Water (H₂O) ≤1.0 %
JP - Identification 1 Passes Test
JP - Identification 2 Passes Test
JP - Water (H₂O) ≤1.0 %
JP - Assay (Anhydrous basis) 97.5 - 102.0 %
JP - Maltose and isomaltose ≤0.4 %
JP - Maltotriose ≤0.2 %
JP - Fructose ≤0.15 %
JP - Unspecified Impurities, each ≤0.10 %
JP - Total Impurities ≤0.5 %
JP - Color and Clarity of Solution Passes Test
JP - Soluble Starch, Sulfites ≤15 ppm
JP - Conductivity (uS cm-1) ≤20 µS/cm
JP - Dextrin Passes Test
JP - Heavy Metals (as Pb) ≤4 ppm
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR