Vacuum systems, chemically resistant, LABOXACT®
Supplier: KNF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SEM820 SEM840 SEM842
189-0025EA 3084 GBP
189-0025 189-0026 189-0027
Vacuum systems, chemically resistant, LABOXACT®
Pumps Vacuum Diaphragm Pumps
These vacuum systems are especially suitable for controlling processes in a rotary evaporator. The patented, closed system arrangement enables gentle distillation and high recovery rates of solvents even with low boiling points.
  • Gentle distillation due to a closed system
  • Reliable control of the evaporation process
  • High solvent recovery
  • Maintenance-free
  • Environmentally friendly
Delivery information: Includes the chemically resistant vacuum pump, base plate, separator, condenser, fine tuning valve, digital vacuum gauge, safety and venting valve,More Product Information