VACUU.LAN® local vacuum network
Supplier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20677101 20677102 20677104 20677105 20677106 20677107 20677155 20677156 20677175 20677176 20677180 20677182 20677190 20677191 20677195 20636668 20677208 20677292 20677293 22614455
181-1489EA 389 GBP
181-1489 181-1490 181-1492 181-1493 181-1494 181-1495 189-0028 181-1499 189-0029 181-1503 181-1505 181-1507 181-1509 181-1510 181-1488 181-1010 181-1019 181-1026 181-1027 181-0484
VACUU.LAN® local vacuum network
Valves Vacuum Valves
The VACUU.LAN local vacuum network connects several workstations to one vacuum pump. The various modules make it possible to connect almost all popular vacuum applications such as rotary evaporators, vacuum concentrators, gel dryers, drying ovens and filter systems. It can be integrated into new laboratory furniture at the time of assembly or subsequently installed in existing laboratories.
  • Individual connection configuration, vacuum connections to suit the requirements of the workstation
  • Integrated non-return valves to prevent cross-contamination, even
More Product Information