Oils for rotary vane pumps
Supplier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20687010 20687011 20687012 20687013
181-1241EA 30.8 GBP
181-1241 181-1242 181-1243 181-0449
Oils for rotary vane pumps
Lubricants
Rotary vane pump oil, B
Rotary vane pump oil, B has excellent viscosity characteristics and low vapour pressure. Its exceptional chemical resistance, more stable behaviour when pumping oxidants, acid and alkaline vapours, and the fact that it is better tolerated on contact with skin, sets this oil apart from conventional mineral oils. Even after continuous operation for several weeks and months, the oil ensures the pressure is maintained constantly. Used for priming all RE/RZ series pumps.
Rotary vane pump oil, K 8
Rotary vane pump oil, K 8 is a special oil that is intended specifically forMore Product Information