Chemistry vacuum pumps and systems, MZ
Supplier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
732801 732500 20732502 732600 20732602 20732700 732800 732300 732501 732601 732301 20732302 724100 20724102
181-0436EA 0 GBP
181-0436 181-1042 181-1043 181-1044 181-1045 181-1046 181-1047 181-1040 181-0163 181-0164 181-0161 181-0338 181-0462 181-0337
Chemistry vacuum pumps and systems, MZ
Pumps Vacuum Diaphragm Pumps
These chemistry vacuum systems can be used in a wide range of applications for evacuating or moving gases and vapours between containers in chemistry and physics laboratories, e.g. as an alternative to water jet pumps and rotary vane pumps. Compact construction, very quiet running, long service life, diaphragm and valves are very easy to replace. Oil-free vacuum.
  • Includes chemistry diaphragm pump with high chemical resistance (parts in contact with media are made from PTFE compounds and similar)
  • High pump volume capacity even at low pressures
  • Standard
More Product Information